missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ STOML2 Recombinant Rabbit Monoclonal Antibody (24GB1870)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557745
This item is not returnable.
View return policy
Description
STOML2 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Mitochondrial protein that probably regulates the biogenesis and the activity of mitochondria. Stimulates cardiolipin biosynthesis, binds cardiolipin-enriched membranes where it recruits and stabilizes some proteins including prohibitin and may therefore act in the organization of functional microdomains in mitochondrial membranes. Through regulation of the mitochondrial function may play a role into several biological processes including cell migration, cell proliferation, T-cell activation, calcium homeostasis and cellular response to stress. May play a role in calcium homeostasis through negative regulation of calcium efflux from mitochondria. Required for mitochondrial hyperfusion a pro-survival cellular response to stress which results in increased ATP production by mitochondria. May also regulate the organization of functional domains at the plasma membrane and play a role in T-cell activation through association with the T-cell receptor signaling complex and its regulation.
Specifications
| STOML2 | |
| Recombinant Monoclonal | |
| 0.7 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q4FZT0, Q99JB2, Q9UJZ1 | |
| Stoml2 | |
| A synthesized peptide derived from human Stomatin like 2 (aa 10-130). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB1870 | |
| Unconjugated | |
| Stoml2 | |
| 0610038F01Rik; EPB72-like 2; EPB72-like protein 2; HSPC108; MSLP2; Paraprotein target 7; paratarg-7; Slp2; SLP-2; stomatin (Epb7.2)-like 2; stomatin (EPB72)-like 2; stomatin like 2; stomatin-like 2; stomatin-like protein 2, mitochondrial; Stoml2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 298203, 30968, 66592 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction