Learn More
STOML1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12862-1-AP-150UL
 View more versions of this product
Product Code. 16860804
  • £349.00 / 150µL
Specifications
STOML1
Polyclonal
Unconjugated
STOML1
EPB72 like protein 1, hUNC 24, Protein unc 24 homolog, SLP 1, SLP1, stomatin (EPB72) like 1, Stomatin like protein 1, Stomatin related protein, STOML1, STORP, UNC24
Rabbit
Antigen Affinity Chromatography
RUO
300748, 69106, 9399
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.23 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8CI66, Q9UBI4
STOML1
STOML1 Fusion Protein Ag3978
150 μL
Primary
Human, Mouse, Rat