missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies STO-609 (Acetate)

Product Code. 17150282
Encompass_Preferred
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
This item is not returnable. View return policy

Product Code. 17150282

Brand: STEMCELL Technologies 73862

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Calcium signaling inhibitor; Inhibits calcium/calmodulin-dependent kinase kinases (CaMKKs)

  • STO-609 is a cell-permeable, selective inhibitor of ATP-competitive calcium/calmodulin-dependent kinase kinases (CaMKK)
  • STO-609 inhibits the activities of recombinant isoforms CaMKKα (Ki = 80 ng/mL) and CaMKKβ (Ki = 15 ng/mL) and also inhibits their autophosphorylation (Kukimoto-Niino et al.; Tokumitsu et al.) STO-609 reduces thrombin-induced nuclear localization of NF-κB activation in endothelial cells (Bair et al.) This product is supplied as the acetate salt form of the molecule
  • Stimulates osteoblast formation and inhibits osteoclast differentiation in mouse mesenchymal stem cells, and reverses age-associated decline in bone mass (Cary et al.; Pritchard et al.)
  • Decreases in vitro growth of human hepatic cancer cell lines, and decreases tumor burden in mouse hepatic tumor model (Lin et al.)

Specifications

CAS 1173022-21-3
Quantity 10 mg
Molecular Formula C19H10N2O3 · C2H4O2
Purity ≥98%
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.