Learn More
STMN2 Mouse anti-Human, Unconjugated, Clone: 1F6C4, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67204-1-IG-150UL
 View more versions of this product
Product Code. 16859715
  • £349.00 / 150µL
Description
The STMN2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human STMN2. This antibody recognizes human, mouse, rat, pig antigen. The STMN2 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
STMN2
Monoclonal
500 μg/mL
Mouse
Primary
Human, Mouse, Rat, Pig
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA
1F6C4
Unconjugated
150 μL
11075
IgG1