Learn More
Invitrogen™ STK40 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA522165
Description
Recommended positive controls: 293T, A431, H1299, HeLa, HepG2, Molt-4, Raji. Predicted reactivity: Mouse (100%), Rat (100%), Xenopus laevis (92%), Chicken (92%), Chimpanzee (100%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
STK40 is a protein coding gene. May be a negative regulator of NF-kappa-B and p53-mediated gene transcription.
Specifications
| STK40 | |
| Polyclonal | |
| Unconjugated | |
| STK40 | |
| 2310004N11Rik; AI428792; Lyk4; ortholog of Ser/Thr-like protein kinase Lyk4; RGD1307310; RP11-268J15.4; Ser/Thr-like kinase; Ser/Thr-like protein kinase lyk4; serine/threonine kinase 40; serine/threonine-protein kinase 40; Serine/threonine-protein kinase lyk4; SgK495; SHIK; SINK-homologous serine/threonine-protein kinase; Stk40; Sugen kinase 495 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 83931 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.97 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| Q8N2I9 | |
| STK40 | |
| Recombinant protein encompassing a sequence within the center region of human STK40. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.