missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ STK40 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA582329
This item is not returnable.
View return policy
Description
Immunogen sequence: AKALGSGISG NNAKRAGPFI LGPRLGNSPV PSIVQCLARK DGTDDFYQLK ILTLEERGDQ GIESQEERQG KMLLHTEYSL LSLLHTQDGV VHHHGLFQDR TCEIVEDTES SRMVKKMKKR ICLVLDCLCA HDFSDKTADL Highest antigen sequence indentity to the following orthologs: Rat - 96%, Mouse - 96%.
STK40 is a protein coding gene. May be a negative regulator of NF-kappa-B and p53-mediated gene transcription.
Specifications
| STK40 | |
| Polyclonal | |
| Unconjugated | |
| STK40 | |
| 2310004N11Rik; AI428792; Lyk4; ortholog of Ser/Thr-like protein kinase Lyk4; RGD1307310; RP11-268J15.4; Ser/Thr-like kinase; Ser/Thr-like protein kinase lyk4; serine/threonine kinase 40; serine/threonine-protein kinase 40; Serine/threonine-protein kinase lyk4; SgK495; SHIK; SINK-homologous serine/threonine-protein kinase; Stk40; Sugen kinase 495 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 83931 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8N2I9 | |
| STK40 | |
| Recombinant protein corresponding to Human STK40. Recombinant protein control fragment (Product #RP-89872). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction