missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ STK40 Polyclonal Antibody
Description
STK40 Polyclonal Antibody for Western Blot, IHC (P), Flow
STK40 is a protein coding gene. May be a negative regulator of NF-kappa-B and p53-mediated gene transcription.
Specifications
Specifications
| Antigen | STK40 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | STK40 |
| Gene Accession No. | Q7TNL3, Q8N2I9 |
| Gene Alias | 2310004N11Rik; AI428792; Lyk4; ortholog of Ser/Thr-like protein kinase Lyk4; RGD1307310; RP11-268J15.4; Ser/Thr-like kinase; Ser/Thr-like protein kinase lyk4; serine/threonine kinase 40; serine/threonine-protein kinase 40; Serine/threonine-protein kinase lyk4; SgK495; SHIK; SINK-homologous serine/threonine-protein kinase; Stk40; Sugen kinase 495 |
| Gene Symbols | STK40 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?