missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ STK4 (MST1) Recombinant Human Protein
STK4 (MST1) is a serine/threonine kinase which may be involved in apoptosis.
Brand: Invitrogen™ PV4177
Code : NEW
Additional Details : Weight : 0.00900kg
Description
It is possible that the STK4 (MST1) protein induces the chromatin condensation observed in the apoptosis process.
Specifications
NP_006273 | |
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol | |
1mg | |
RUO - research use only | |
STK4 (MST1) | |
Human | |
GST Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
83.1kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
STE20 Kinase Family | |
Recombinant | |
Baculovirus |