Learn More
Invitrogen™ Human STK39 (STLK3), His Tag Recombinant Protein
Description
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Recombinant full-length human STK39 (STLK3) was expressed using baculovirus in Sf9 insect cells using an N-terminal His tag. , Validated in LanthaScreen™ Kinase Binding Assay, Protocols available for simple use and optimized performance, Multiple pack sizes available for convenient use Protein Form: Recombinant The gene accession number for STK39 (STLK3) is NP_037365.2.
Specifications
Specifications
| Accession Number | Q9UEW8 |
| Concentration | See Label |
| For Use With (Application) | Kinase Assay |
| Formulation | 50 mM sodium phosphate with 0.1 mM PMSF, 0.2 mM DTT, 150 mM Imidazole, 25% glycerol, 300 mM NaCl and no preservative; pH 7 |
| Gene ID (Entrez) | 27347 |
| Molecular Weight (g/mol) | 60.6 kDa |
| Name | Human STK39 (STLK3), His Tag |
| Quantity | 10 μg |
| Gene Alias | AW227544; AW556857; DCHT; DKFZp686K05124; kidney-specific SPAK; pancreatic serine/threonine-protein kinase; Pask; proline-alanine-rich STE20-related kinase; PS/TK; PSTK1; RF005; Rnl5; serine threonine kinase 39; serine threonine kinase 39 (STE20/SPS1 homolog, yeast); serine/threonine kinase 39; serine/threonine kinase 39, STE20/SPS1 homolog; serine/threonine-protein kinase 39; small intestine SPAK-like kinase; Spak; Ste-20 related kinase; STE20/SPS1 homolog; STE20/SPS1-related proline-alanine-rich protein kinase; Ste20-like protein kinase; ste-20-related kinase; Stk39 |
| Common Name | STK39 |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.