missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STK39 Antibody (OTI4E3), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-48021
This item is not returnable.
View return policy
Description
STK39 Monoclonal specifically detects STK39 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| STK39 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1000, Flow Cytometry 1:100, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:50, Immunohistochemistry-Paraffin 1:50 | |
| STK39 | |
| Full length human recombinant protein of human STK39 (NP_037365) produced in HEK293T cell. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This antibody is specific for Homo sapiens serine threonine kinase 39 (STE20/SPS1 homolog, yeast) (STK39). | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| OTI4E3 | |
| Unconjugated | |
| DCHTsmall intestine SPAK-like kinase, DKFZp686K05124, EC 2.7.11, EC 2.7.11.1, PASK, proline-alanine-rich STE20-related kinase, serine threonine kinase 39, Serine/threonine-protein kinase 39, SPAKserine threonine kinase 39 (STE20/SPS1 homolog, yeast), STE20/SPS1 homolog, STE20/SPS1-related proline-alanine-rich protein kinase, Ste20-like protein kinase, Ste-20-related kinase | |
| Mouse | |
| 59.3 kDa | |
| 0.1 mL | |
| Protein Kinase | |
| 27347 | |
| Human, Mouse, Rat | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction