missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STK39 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-51955
This item is not returnable.
View return policy
Description
STK39 Polyclonal antibody specifically detects STK39 in Human samples. It is validated for Western Blot, ELISA.
Specifications
| STK39 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| DCHTsmall intestine SPAK-like kinase, DKFZp686K05124, EC 2.7.11, EC 2.7.11.1, PASK, proline-alanine-rich STE20-related kinase, serine threonine kinase 39, Serine/threonine-protein kinase 39, SPAKserine threonine kinase 39 (STE20/SPS1 homolog, yeast), STE20/SPS1 homolog, STE20/SPS1-related proline-alanine-rich protein kinase, Ste20-like protein kinase, Ste-20-related kinase | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Cross reacts with Human. Expected from sequence similarity: Mouse, Rat and Dog. | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot, ELISA, Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| Western Blot 0.3 - 1 μg/mL, Peptide ELISA Detection limit 1:4000 | |
| NP_037365.2 | |
| STK39 | |
| Peptide with sequence C-SQEKSRRVKEENPE corresponding to internal region according to NP_037365.2. | |
| 0.1 mg | |
| Protein Kinase | |
| 27347 | |
| Human, Mouse, Rat | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu