missing translation for 'onlineSavingsMsg'
Learn More

STK38L Antibody (OTI4D8), DyLight 594, Novus Biologicals™

Product Code. 18416885 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18416885 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18416885 Supplier Novus Biologicals Supplier No. NBP274403DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

STK38L Monoclonal antibody specifically detects STK38L in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen STK38L
Applications Western Blot, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI4D8
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias EC 2.7.11, EC 2.7.11.1, KIAA0965nuclear Dbf2-related 2, NDR2 protein kinase, NDR2serine/threonine-protein kinase 38-like, Nuclear Dbf2-related kinase 2, serine/threonine kinase 38 like
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 1-281 of human STK38L (NP_055815) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 23012
Target Species Human, Mouse, Rat, Canine, Monkey
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.