missing translation for 'onlineSavingsMsg'
Learn More

STK38L Antibody (4E5), Novus Biologicals™

Product Code. 18365968 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18365968 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18365968 Supplier Novus Biologicals Supplier No. H00023012M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

STK38L Monoclonal antibody specifically detects STK38L in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen STK38L
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 4E5
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH28603
Gene Alias EC 2.7.11, EC 2.7.11.1, KIAA0965nuclear Dbf2-related 2, NDR2 protein kinase, NDR2serine/threonine-protein kinase 38-like, Nuclear Dbf2-related kinase 2, serine/threonine kinase 38 like
Host Species Mouse
Immunogen STK38L (AAH28603, 1 a.a. ~ 464 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MAMTAGTTTTFPMSNHTRERVTVAKLTLENFYSNLILQHEERETRQKKLEVAMEEEGLADEEKKLRRSQHARKETEFLRLKRTRLGLDDFESLKVIGRGAFGEVRLVQKKDTGHIYAMKILRKSDMLEKEQVAHIRAERDILVEADGAWVVKMFYSFQDKRNLYLIMEFLPGGDMMTLLMKKDTLTEEETQFYISETVLAIDAIHQLGFIHRDIKPDNLLLDAKGHVKLSDFGLCTGLKKAHRTEFYRNLTHNPPSDFSFQNMNSKRKAETWKKNRRQLAYSTVGTPDYIAPEVFMQTGYNKLCDWWSLGVIMYEMLIGYPPFCSETPQETYRKVMNWKETLVFPPEVPISEKAKDLILRFCIDSENRIGNSGVEEIKGHPFFEGVDWEHIRERPAAIPIEIKSIDDTSNFDDFPESDILQPVPNTTEPDYKSKDWVFLNYTYKRFEGLTQRGSIPTYMKAGKL
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 23012
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.