Learn More
Invitrogen™ Human STK38 (NDR), GST Tag Recombinant Protein
Description
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Full-length recombinant human STK38 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. , Validated in LanthaScreen™ Kinase Binding Assay, Protocols available for simple use and optimized performance, Multiple pack sizes available for convenient use Protein Form: Recombinant The gene accession number for STK38 (NDR) is NP_009202.1.
Specifications
Specifications
| Accession Number | Q15208 |
| Concentration | See Label |
| For Use With (Application) | Kinase Assay |
| Formulation | 50 mM tris HCl with 0.1 mM EDTA, 0.1 mM PMSF, 0.25 mM DTT, 10 mM glutathione, 150 mM NaCl, 25% glycerol and no preservative; pH 7.5 |
| Gene ID (Entrez) | 11329 |
| Molecular Weight (g/mol) | 80.5 kDa |
| Name | Human STK38 (NDR), GST Tag |
| Quantity | 1 mg |
| Gene Alias | 5830476G13Rik; 9530097A09Rik; AA617404; NDR; Ndr Ser/Thr kinase-like protein; NDR1; NDR1 protein kinase; nuclear Dbf2-related kinase 1; serine/threonine kinase 38; serine/threonine-protein kinase 38; STK38 |
| Common Name | STK38 |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.