missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STK36 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12559-1-AP-150UL
This item is not returnable.
View return policy
Description
Serine/threonine protein kinase required for postnatal development, possibly by regulating the homeostasis of cerebral spinal fluid or ciliary function. Controls the activity of the transcriptional regulators GLI1, GLI2 and GLI3 by opposing the effect of SUFU and promoting their nuclear localization. GLI2 requires an additional function of STK36 to become transcriptionally active, but the enzyme does not need to possess an active kinase catalytic site for this to occur.Specifications
| STK36 | |
| Polyclonal | |
| Unconjugated | |
| STK36 | |
| FU, Fused homolog, KIAA1278, STK36 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 269209, 27148, 301516 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.15 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q69ZM6, Q9NRP7 | |
| STK36 | |
| STK36 Fusion Protein Ag3279 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction