missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ STK35 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565001
This item is not returnable.
View return policy
Description
Immunogen sequence: TSLKRRHQNVV QFEECVLQRN GLAQRMSHGN KSSQLYLRLV ETSLKGERIL GYAEEPCYLW F Highest antigen sequence identity to the following orthologs - mouse 97%, rat 97%.
The protein encoded by this gene is a kinase that is predominantly found in the nucleus. However, it can interact with PDLIM1/CLP-36 in the cytoplasm and localize to actin stress fibers. The encoded protein may be a regulator of actin stress fibers in nonmuscle cells.
Specifications
| STK35 | |
| Polyclonal | |
| Unconjugated | |
| Stk35 | |
| 1700054C12Rik; bA550O8.2; bA550O8.2 CLIK1; CLIK1; CLIK-1; CLP-36 interacting kinase; CLP-36-interacting kinase 1; OTTHUMP00000030006; PDIK1; PDLIM1-interacting kinase 1; serine threonine kinase 35 long form; serine/threonine kinase 35; serine/threonine-protein kinase 35; Serine/threonine-protein kinase 35 L1; STK35; Stk35l1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 140901 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8TDR2 | |
| Stk35 | |
| Recombinant Human STK35. Recombinant protein control fragment (Product #RP-105766). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction