missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STK35 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£157.00 - £364.00
Specifications
| Antigen | STK35 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
STK35 Polyclonal antibody specifically detects STK35 in Mouse samples. It is validated for Western BlotSpecifications
| STK35 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Protein Kinase | |
| PBS (pH 7.3), 50% glycerol | |
| 140901 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse | |
| bA550O8.2, CLIK-1, CLIK1Serine/threonine-protein kinase 35 L1, CLP-36 interacting kinase, CLP-36-interacting kinase 1, EC 2.7.11, EC 2.7.11.1, PDIK1, PDLIM1-interacting kinase 1, serine threonine kinase 35 long form, serine/threonine kinase 35, serine/threonine-protein kinase 35, STK35L1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 190-380 of human STK35 (NP_543026.2). RPEGGGGSARPRYSLLAEIGRGSYGVVYEAVAGRSGARVAVKKIRCDAPENVELALAEFWALTSLKRRHQNVVQFEECVLQRNGLAQRMSHGNKSSQLYLRLVETSLKGERILGYAEEPCYLWFVMEFCEGGDLNQYVLSRRPDPATNKSFMLQLTSAIAFLHKNHIVHRDLKPDNILITERSGTPILKVA | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title