Learn More
Invitrogen™ Human STK32C (YANK3), GST Tag Recombinant Protein
Description
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Recombinant human STK32C (YANK3) (66-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. , Validated in LanthaScreen™ Kinase Binding Assay, Protocols available for simple use and optimized performance, Multiple pack sizes available for convenient use Protein Form: Recombinant The gene accession number for STK32C (YANK3) is NP_775846.2.
Specifications
Specifications
| Accession Number | Q86UX6 |
| Concentration | See Label |
| For Use With (Application) | Kinase Assay |
| Formulation | 50 mM tris HCl with 0.1 mM EDTA, 0.1 mM PMSF, 0.25 mM DTT, 10 mM glutathione, 150 mM NaCl, 25% glycerol and no preservative; pH 7.5 |
| Gene ID (Entrez) | 282974 |
| Molecular Weight (g/mol) | 74.6 kDa |
| Name | Human STK32C (YANK3), GST Tag |
| Quantity | 100 μg |
| Gene Alias | MNCb-1563; Pk.; Pk. protein kinase; Pkek; RP11-140A10.1; serine/threonine kinase 32 C; serine/threonine-protein kinase 32 C; STK32C; testicular tissue protein Li 187; YANK3; Yet another novel kinase 3 |
| Common Name | STK32C |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.