missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STK32C Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11434-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a member of the serine/threonine protein kinase family. The specific function of this kinase is not known.Specifications
| STK32C | |
| Polyclonal | |
| Unconjugated | |
| STK32C | |
| PKE, RP11 140A10.1, serine/threonine kinase 32C, STK32C, YANK3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 282974, 365381, 57740 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q86UX6, Q8QZV4 | |
| STK32C | |
| STK32C Fusion Protein Ag1960 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction