missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STK32B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15438-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a serine-threonine protein kinase. Serine-threonine kinases transfer phosphate molecules to the oxygen atoms of serine and threonine. A genomic deletion affecting this gene has been associated with Ellis-van Creveld syndrome, an autosomal recessive skeletal dysplasia. Alternative splicing results in multiple transcript variants.Specifications
| STK32B | |
| Polyclonal | |
| Unconjugated | |
| STK32B | |
| HSA250839, serine/threonine kinase 32B, STK32, STK32B, STKG6, YANK2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 305431, 55351, 64293 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.15 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9JJX8, Q9NY57 | |
| STK32B | |
| STK32B Fusion Protein Ag4507 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction