Learn More
STK32B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 15438-1-AP-150UL
 View more versions of this product
Product Code. 16804614
  • £349.00 / 150µL
Description
This gene encodes a serine-threonine protein kinase. Serine-threonine kinases transfer phosphate molecules to the oxygen atoms of serine and threonine. A genomic deletion affecting this gene has been associated with Ellis-van Creveld syndrome, an autosomal recessive skeletal dysplasia. Alternative splicing results in multiple transcript variants.
Specifications
Specifications
STK32B
Polyclonal
Unconjugated
STK32B
HSA250839, serine/threonine kinase 32B, STK32, STK32B, STKG6, YANK2
Rabbit
Antigen Affinity Chromatography
RUO
305431, 55351, 64293
-20°C
Liquid
Western Blot
0.15 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9JJX8, Q9NY57
STK32B
STK32B Fusion Protein Ag4507
150 μL
Primary
Human, Mouse, Rat