missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ STK32B Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566178
This item is not returnable.
View return policy
Description
Immunogen sequence: LRKLLTKDPES RVSSLHDIQS VPYLADMNWD AVFKKALMPG FV Highest antigen sequence identity to the following orthologs - mouse 86%, rat 84%.
This gene encodes a serine-threonine protein kinase. Serine-threonine kinases transfer phosphate molecules to the oxygen atoms of serine and threonine. A genomic deletion affecting this gene has been associated with Ellis-van Creveld syndrome, an autosomal recessive skeletal dysplasia. Alternative splicing results in multiple transcript variants.
Specifications
| STK32B | |
| Polyclonal | |
| Unconjugated | |
| STK32B | |
| 2510009F08Rik; gene for serine/threonine protein kinase; HSA250839; serine threonine kinase 32; serine/threonine kinase 32B; serine/threonine-protein kinase 32B; Stk32; Stk32b; STKG6; UNQ3003/PRO9744; YANK2; Yet another novel kinase 2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55351 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NY57 | |
| STK32B | |
| Recombinant Human STK32B. Recombinant protein control fragment (Product #RP-106866). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction