missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STK32A Antibody (3E8-1D3), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00202374-M01
This item is not returnable.
View return policy
Description
STK32A Monoclonal antibody specifically detects STK32A in Human samples. It is validated for Western Blot, ELISA, KnockDown
Specifications
| STK32A | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| A930015B13Rik, EC 2.7.11, EC 2.7.11.1, MGC22688, serine/threonine kinase 32A, YANK1serine/threonine-protein kinase 32A | |
| STK32A (AAH21666, 1 a.a. ∽ 166 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MGANTSRKPPVFDENEDVNFDHFEILRAIGKGSFGKVCIVQKNDTKKMYAMKYMNKQKCVERNEVRNVFKELQIMQGLEHPFLVNLWYSFQDEEDMFMVVDLLLGGDLRYHLQQNVHFKEETVKLFICELVMALDYLQNQRIIHRDMKPDNILLDEHDTWLSYKSH | |
| 0.1 mg | |
| Protein Kinase | |
| 202374 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Western Blot, ELISA, KnockDown | |
| 3E8-1D3 | |
| Western Blot 1:500, ELISA, Knockdown Validated | |
| AAH21666 | |
| Mouse | |
| IgG purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction