missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STK31 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14140-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene is similar to a mouse gene that encodes a putative protein kinase with a tudor domain, and shows testis-specific expression. Alternative splicing results in multiple transcript variants encoding different isoforms.Specifications
| STK31 | |
| Polyclonal | |
| Unconjugated | |
| STK31 | |
| serine/threonine kinase 31, SGK396, STK31, Sugen kinase 396, TDRD8 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 500120, 56164, 77485 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.35 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q99MW1, Q9BXU1 | |
| STK31 | |
| STK31 Fusion Protein Ag5221 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction