missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ STK29 Polyclonal Antibody

Description
A suggested positive control is human brain tissue lysate. PA5-20358 can be used with blocking peptide PEP-0478.
BRSK2 is a brain-selective serine/threonine kinase 2 that is mainly expressed in the brain, with weaker expression in testis and pancreas. BRSK2 expressed in insect cells specifically phosphorylates WEE1A, CDC25C and CDC25B in an in vitro assay. DNA damage induced by ultraviolet (UV) irradiation or methyl methane sulfonate, but not by ionizing radiation, enhanced endogenous BRSK2 kinase activity in a caffeine-sensitive manner and caused translocation of BRSK2 from the cytoplasm to the nucleus. Overexpression of BRSK2 in HeLa cells results in increased phosphorylation of CDC25C. DNA damage induced by ultraviolet (UV) irradiation or methyl methane sulfonate, but not by ionizing radiation, enhances endogenous BRSK2 kinase activity in a caffeine-sensitive manner and causes translocation of BRSK2 from the cytoplasm to the nucleus. Overexpression of BRSK2 induces G2/M arrest in HeLa cells. Small interfering RNA against BRSK2 partly abrogates UV-induced G2/M arrest. BRSK2 may act as a checkpoint kinase upon DNA damage induced by UV irradiation or methyl methane sulfonate.
Specifications
Specifications
| Antigen | STK29 |
| Applications | Immunohistochemistry, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | Brsk2 |
| Gene Accession No. | Q69Z98, Q8IWQ3 |
| Gene Alias | 4833424K13Rik; BR serine/threonine kinase 2; BR serine/threonine-protein kinase 2; brain serine/threonine kinase 2; brain-selective kinase 2; Brain-specific serine/threonine-protein kinase 2; BRSK2; C11orf7; HUSSY-12; Kiaa4256; PEN11B; protein kinase SAD1B; SAD1; SAD1B; Sada; SAD-A; serine/threonine-protein kinase 29; Serine/threonine-protein kinase BRSK2; serine/threonine-protein kinase SAD-A; STK29; testicular tissue protein Li 30 |
| Gene Symbols | Brsk2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?