missing translation for 'onlineSavingsMsg'
Learn More

STK25 Antibody (4B10), Novus Biologicals™

Product Code. 18337328 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18337328 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18337328 Supplier Novus Biologicals Supplier No. H00010494M02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

STK25 Monoclonal antibody specifically detects STK25 in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen STK25
Applications Western Blot, ELISA, Immunoprecipitation
Classification Monoclonal
Clone 4B10
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH07852
Gene Alias EC 2.7.11, EC 2.7.11.1, serine/threonine kinase 25, serine/threonine kinase 25 (Ste20, yeast homolog), serine/threonine-protein kinase 25, SOK-1, SOK1DKFZp686J1430, Ste20, yeast homolog, Ste20/oxidant stress response kinase 1, sterile 20 (oxidant stress response kinase 1, yeast Sps1/Ste20-related kinase1), Sterile 20/oxidant stress-response kinase 1, YSK1yeast)
Host Species Mouse
Immunogen STK25 (AAH07852, 321 a.a. ~ 426 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. FPPTIRPSPHSKLHKGTALHSSQKPAEPVKRQPRSQCLSTLVRPVFGELKEKHKQSGGSVGALEELENAFSLAEESCPGISDKLMVHLVERVQRFSHNRNHLTSTR
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 10494
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG3 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.