missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human STK22D (TSSK1), His Tag Recombinant Protein

Product Code. 10492303
Change view
Click to view available options
Quantity:
1 mg
100 μg
Unit Size:
100µg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10492303 1 mg 1mg
15468433 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10492303 Supplier Invitrogen™ Supplier No. PV4063

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Recombinant, Full Length

STK22D (TSSK1) is a serine/threonine kinase essential to the function and production of sperm (spermiogenesis). TSSK1 belongs to a family of serine/threonine kinases highly expressed in testis.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9BXA7
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50 mM tris HCl with 0.05% NP-40, 0.5 mM EDTA, 100 mM NaCl, 4 mM DTT, 50% glycerol and no preservative; pH 7.5
Gene ID (Entrez) 83942
Molecular Weight (g/mol) 46.7 kDa
Name Human STK22D (TSSK1), His Tag
Quantity 1 mg
Regulatory Status RUO - research use only
Gene Alias FKSG81; serine/threonine kinase 22 A (spermiogenesis associated); serine/threonine kinase 22 D (spermiogenesis associated); serine/threonine kinase FKSG81; serine/threonine-protein kinase 22 A; spermiogenesis associated; spermiogenesis associated 4; SPOGA1; SPOGA4; Stk22a; STK22D; testis specific serine kinase 1 B; testis tissue sperm-binding protein Li 55 e; testis-specific kinase 1; testis-specific serine kinase 1; testis-specific serine kinase 1 B; testis-specific serine/threonine kinase; testis-specific serine/threonine-protein kinase 1; TSK1; TSK-1; Tssk; TSSK1; TSSK-1; TSSK1B
Common Name TSSK1
KinaseFamily TSSK Kinase Family
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-tag
Sequence Full length
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Expression System Insect cells
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.