missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STK22C Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38448-100ul
This item is not returnable.
View return policy
Description
STK22C Polyclonal antibody specifically detects STK22C in Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| STK22C | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| EC 2.7.11, EC 2.7.11.1, serine/threonine kinase 22C (spermiogenesis associated), Serine/threonine-protein kinase 22C, SPOGA3TSK3, STK22Cspermiogenesis associated 3, STK22D, Testis-specific kinase 3, testis-specific serine kinase 3, testis-specific serine/threonine kinase 22C, testis-specific serine/threonine-protein kinase 3, TSK-3, TSSK-3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-268 of human STK22C (NP_443073.1).,, Sequence:, MEDFLLSNGYQLGKTIGEGTYSKVKEAFSKKHQRKVAIKVIDKMGGPEEFIQRFLPRELQIVRTLDHKNIIQVYEMLESADGKICLVMELAEGGDVFDCVLNGGPLPESRAKALFRQMVEAIRYCHGCGVAHRDLKCENALLQGFNLKLTDFGFAKVLPKSHRELSQTFCGSTAYAAPEVLQGIPHDSKKGDVWSMGVVLYVMLCASLPFDDTDIPKMLWQQQKGVSFPTHLSISADCQDLLKRLLEPDMILRPSIEEVSWHPWLAST | |
| 100 μL | |
| Protein Kinase | |
| 81629 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction