missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STK16 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99954-50ul
This item is not returnable.
View return policy
Description
STK16 Polyclonal antibody specifically detects STK16 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| STK16 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:500-1:2000 | |
| EC 2.7.11.1, FLJ39635, hPSK, MGC16211, MPSK1, MPSKFLJ39635, Myristoylated and palmitoylated serine/threonine-protein kinase, PKL12KRCT, protein kinase expressed in day 12 fetal liver, Protein kinase PKL12, serine/threonine kinase 16, serine/threonine-protein kinase 16, TGF-beta-stimulated factor 1, TSF1, TSF-1 | |
| Produced in rabbits immunized with purified, recombinant Human STK16 (Accession#: AAH02618.1; Met1-Ile305) | |
| 50 μg | |
| Signal Transduction | |
| 8576 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction