missing translation for 'onlineSavingsMsg'
Learn More

STK11IP Antibody, Alexa Fluor™ 405, Novus Biologicals™

Product Code. 18427023 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18427023 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18427023 Supplier Novus Biologicals Supplier No. NBP130109AF405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

STK11IP Polyclonal antibody specifically detects STK11IP in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen STK11IP
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 405
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias KIAA1898LKB1 interacting protein, LIP1serine/threonine kinase 11-interacting protein, LKB1-interacting protein 1, LKB1IPserine/threonine-protein kinase 11-interacting protein, serine/threonine kinase 11 interacting protein, STK11IP1STK11 interacting protein
Host Species Rabbit
Immunogen maps to a region between residue 400 and 450 of human serine/threonine kinase 11 interacting protein using the numbering given in entry NP_443134.2
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114790
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.