Learn More
Heidolph™ Stirrer Guide (NS 29/32 PTFE)
Suitable for vacuum, perfect bearing for stainless steel and glass stirrer shafts
Brand: Heidolph™ 509-09000-00
Additional Details : Weight : 0.01000kg
Description
- Ground joint cone made of PTFE with sealing rings on the outside to prevent sticking of the connection and to reduce danger of breaking glass
- A special gasket made of PTFE and an FPM o-ring which is compressed by a GL screw cap provides good sealing of the stirrer shaft
- Accepts o.d. 8 mm shafts
Specifications
Suitable for vacuum, perfect bearing for stainless steel and glass stirrer shafts | |
Stirrer Guide | |
PTFE |