missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ STIP1 Recombinant Rabbit Monoclonal Antibody (3E4)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA533200

Product Code. 16565831

  • £469.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

STIP1 Recombinant Monoclonal Antibody for Western Blot, ELISA

Mediates the association of the molecular chaperones HSC70 and HSP90 (HSPCA and HSPCB).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

STIP1
Recombinant Monoclonal
1.2 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P31948
STIP1
A synthesized peptide derived from human STIP1.
100 μL
Primary
Human
Antibody
IgG
ELISA, Western Blot
3E4
Unconjugated
STIP1
epididymis secretory sperm binding protein Li 94n; HEL-S-94n; HOP; hsc70/Hsp90-organizing protein; Hsp70/Hsp90 organizing protein; Hsp70/Hsp90-organizing protein; IEF SSP 3521; IEF-SSP-3521; mSTI1; NY-REN-11 antigen; P60; Renal carcinoma antigen NY-REN-11; STI1; STI1L; Stip1; stress induced phosphoprotein 1; stress-induced phosphoprotein 1; stress-induced-phosphoprotein 1; stress-induced-phosphoprotein 1 (Hsp70/Hsp90-organizing protein); stress-inducible protein; transformation-sensitive protein IEF SSP 3521
Rabbit
Antigen Affinity Chromatography, Protein G
RUO
10963
-20°C or -80°C if preferred
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.