missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STIP1 Polyclonal Antibody

STIP1 Polyclonal Antibody
Brand: Invitrogen PA517193
This item is not returnable.
View return policy
Description
STIP1 Polyclonal Antibody for Western Blot
Specifications
| STIP1 | |
| Polyclonal | |
| 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5 | |
| O35814, P31948, Q60864 | |
| STIP1 | |
| Synthetic peptide corresponding to human Hop. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| STIP1 | |
| epididymis secretory sperm binding protein Li 94n; HEL-S-94n; HOP; hsc70/Hsp90-organizing protein; Hsp70/Hsp90 organizing protein; Hsp70/Hsp90-organizing protein; IEF SSP 3521; IEF-SSP-3521; mSTI1; NY-REN-11 antigen; P60; Renal carcinoma antigen NY-REN-11; STI1; STI1L; Stip1; stress induced phosphoprotein 1; stress-induced phosphoprotein 1; stress-induced-phosphoprotein 1; stress-induced-phosphoprotein 1 (Hsp70/Hsp90-organizing protein); stress-inducible protein; transformation-sensitive protein IEF SSP 3521 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10963, 192277, 20867 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction