missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ STIP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106874
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total STIP1.
Mediates the association of the molecular chaperones HSC70 and HSP90 (HSPCA and HSPCB).
Specifications
| STIP1 | |
| Polyclonal | |
| Unconjugated | |
| STIP1 | |
| epididymis secretory sperm binding protein Li 94n; HEL-S-94n; HOP; hsc70/Hsp90-organizing protein; Hsp70/Hsp90 organizing protein; Hsp70/Hsp90-organizing protein; IEF SSP 3521; IEF-SSP-3521; mSTI1; NY-REN-11 antigen; P60; Renal carcinoma antigen NY-REN-11; STI1; STI1L; Stip1; stress induced phosphoprotein 1; stress-induced phosphoprotein 1; stress-induced-phosphoprotein 1; stress-induced-phosphoprotein 1 (Hsp70/Hsp90-organizing protein); stress-inducible protein; transformation-sensitive protein IEF SSP 3521 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 10963, 192277, 20867 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O35814, P31948, Q60864 | |
| STIP1 | |
| A synthesized peptide derived from human STIP1(Accession P31948), corresponding to amino acid residues L7-A57. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction