missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STING/TMEM173 Antibody (STING1/8187R), Alexa Fluor™ 700, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20699AF700
This item is not returnable.
View return policy
Description
STING/TMEM173 Monoclonal antibody specifically detects STING/TMEM173 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| STING/TMEM173 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| endoplasmic reticulum IFN stimulator, Endoplasmic reticulum interferon stimulator, ERIS, FLJ38577, hMITA, hSTING, Mediator of IRF3 activation, MITA, mitochondrial mediator of IRF3 activation, MPYS, NET23, N-terminal methionine-proline-tyrosine-serine plasma membrane tetraspanner, SAVI, Stimulator of interferon genes protein, stimulator of interferon protein, sting, STING-beta, TMEM173, transmembrane protein 173 | |
| Recombinant fragment (around aa190-290) of human STING/TMEM173 protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Cancer, Immunology, Innate Immunity | |
| 340061 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| STING1/8187R | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction