missing translation for 'onlineSavingsMsg'
Learn More

STING/TMEM173 Antibody (STING1/7431), DyLight 350, Novus Biologicals™

Product Code. 18386542
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18386542 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18386542 Supplier Bio-Techne Supplier No. NBP314076UV

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

STING/TMEM173 Monoclonal antibody specifically detects STING/TMEM173 in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen STING/TMEM173
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone STING1/7431
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias endoplasmic reticulum IFN stimulator, Endoplasmic reticulum interferon stimulator, ERIS, FLJ38577, hMITA, hSTING, Mediator of IRF3 activation, MITA, mitochondrial mediator of IRF3 activation, MPYS, NET23, N-terminal methionine-proline-tyrosine-serine plasma membrane tetraspanner, SAVI, Stimulator of interferon genes protein, stimulator of interferon protein, sting, STING-beta, TMEM173, transmembrane protein 173
Host Species Mouse
Immunogen Recombinant fragment (around aa190-290) of human STING/TMEM173 protein (exact sequence is proprietary) (Uniprot: Q86WV6 )
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 340061
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.