missing translation for 'onlineSavingsMsg'
Learn More

STING/TMEM173 Antibody, Janelia Fluor™ 549, Novus Biologicals™

Product Code. 18415246 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18415246 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18415246 Supplier Novus Biologicals Supplier No. NBP224683JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

STING/TMEM173 Polyclonal antibody specifically detects STING/TMEM173 in Human, Mouse, Canine, Primate, Macaque samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
TRUSTED_SUSTAINABILITY

Specifications

Antigen STING/TMEM173
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias endoplasmic reticulum IFN stimulator, Endoplasmic reticulum interferon stimulator, ERIS, FLJ38577, hMITA, hSTING, Mediator of IRF3 activation, MITA, mitochondrial mediator of IRF3 activation, MPYS, NET23, N-terminal methionine-proline-tyrosine-serine plasma membrane tetraspanner, SAVI, Stimulator of interferon genes protein, stimulator of interferon protein, sting, STING-beta, TMEM173, transmembrane protein 173
Host Species Rabbit
Immunogen Partial synthetic peptide made to an internal portion of human STING/TMEM173 (between amino acids 310-360) [UniProt Q86WV6]
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 340061
Target Species Human, Mouse, Canine, Primate, Macaque
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.