missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STING/TMEM173 Antibody (723505) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB7169JF585
This item is not returnable.
View return policy
Description
STING/TMEM173 Monoclonal antibody specifically detects STING/TMEM173 in Human samples. It is validated for Immunoprecipitation,Western Blot,Flow Cytometry (Intracellular Staining),Immunocytochemistry,CyTOF
Specifications
| STING/TMEM173 | |
| Monoclonal | |
| Janelia Fluor 585 | |
| endoplasmic reticulum IFN stimulator, Endoplasmic reticulum interferon stimulator, ERIS, FLJ38577, hMITA, hSTING, Mediator of IRF3 activation, MITA, mitochondrial mediator of IRF3 activation, MPYS, NET23, N-terminal methionine-proline-tyrosine-serine plasma membrane tetraspanner, SAVI, Stimulator of interferon genes protein, stimulator of interferon protein, sting, STING-beta, TMEM173, transmembrane protein 173 | |
| E. coli-derived recombinant human STING/TMEM173, Ala215-Ser379, Accession # Q86WV6 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Immunoprecipitation, Western Blot, Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF | |
| 723505 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Cancer, Immunology, Innate Immunity | |
| 340061 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction