Learn More
Invitrogen™ STING Recombinant Rabbit Monoclonal Antibody (23GB2320), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA552501
 View more versions of this product
Product Code. 30089448
  • £162.00 / 20µL
Description
STING is a 379 amino acid containing signaling protein that acts as an important component of innate immune signaling. This signaling protein is able to activate both NF-kappa-B and IRF3 transcription pathways to induce expression of type I interferon (IFN-alpha and IFN-beta) and exert a potent anti-viral state following expression. STING may have translocon function, the translocon possibly being able to influence the induction of type I interferons and is also involved in transduction of apoptotic signals via its association with the major histocompatibility complex class II (MHC-II) thus facilitating the detection of intracellular viral RNA species as well as B-form DNA and mediates death signaling via activation of the extracellular signal-regulated kinase (ERK) pathway. Generally seen in endoplasmic reticulum membrane, cell membrane and mitochondrion outer membrane, it is ubiquitously expressed in most of tissues.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
STING
Recombinant Monoclonal
Unconjugated
TMEM173
2610307O08Rik; endoplasmic reticulum IFN stimulator; endoplasmic reticulum interferon stimulator; ERIS; hMITA; hSTING; hypothetical LOC533661; mediator of IRF3 activation; Mita; mitochondrial mediator of IRF3 activation; mitochondrial transmembrane protein 173; MMITA; Mpys; mSTING; NET23; N-terminal methionine-proline-tyrosine-serine plasma membrane tetraspanner; poSTING; RGD1562552; rSTING; SAVI; Stimulator of interferon genes protein; stimulator of interferon response cGAMP interactor 1; STING; STING1; tm173; Tmem173; Transmembrane protein 173
Rabbit
Affinity chromatography
RUO
340061
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
23GB2320
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q86WV6
TMEM173
A synthesized peptide derived from human TMEM173 (250-379AA).
20 μL