missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ STING Degrader SP23
GREENER_CHOICE

Product Code. 30000414
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
This item is not returnable. View return policy

Product Code. 30000414

Brand: Tocris Bioscience™ 8053/5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective STING Degrader (PROTAC™)

STING Degrader SP23 is a selective stimulator of interferon gene (STING) Degrader (PROTAC™; DC50 = 3.2 μM). Comprises the STING inhibitor (C-170), joined by a linker to a CRBN ligand Pomalidomide (Cat. No. 6302). It does not degrade JAK2, STAT3, PI3K, AKT and TLR4 in THP-1 cells. STING Degrader SP23 dose dependently reduces the levels of IFN-β, IL-6, and CXCL10 triggered by cGAMP in THP-1 cells; it also modulates the downstream signals of the STING signaling pathway in vivo. STING Degrader SP23 has high anti-inflammatory efficacy in a cisplatin-induced acute kidney injury mouse model.

Negative control STING Degrader N-Me-SP23 (Cat. No. 8054) also available.

PROTAC™ is a registered trademark of Arvinas Operations, Inc., and is used under license.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2E)-N1-[6-[[2-(2,6-Dioxo-3-piperidinyl)-2,3-dihydro-1,3-dioxo-1H-isoindol-4-yl]amino]hexyl]-N4-[4-[[(5-nitro-2-furanyl)carbonyl]amino]phenyl]-2-butenediamide
CAS 2762552-74-7
Quantity 5 mg
Target Active Degraders
Molecular Formula C34H33N7O10
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.