missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ STING Degrader N-Me-SP23

Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Description
STING Degrader N-Me-SP23 is a negative control for STING Degrader SP23 (Cat. No. 8053).
Specifications
Specifications
| Chemical Name or Material | (2E)-N1-[6-[[2,3-Dihydro-2-(1-methyl-2,6-dioxo-3-piperidinyl)-1,3-dioxo-1H-isoindol-4-yl]amino]hexyl]-N4-[4-[[(5-nitro-2-furanyl)carbonyl]amino]phenyl]-2-butenediamide |
| CAS | 2767427-87-0 |
| Quantity | 5 mg |
| Target | Active Degrader Negative Controls |
| Molecular Formula | C35H35N7O10 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction