missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ STING agonist C53
GREENER_CHOICE

Product Code. 18717306
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18717306

Brand: Tocris Bioscience™ 7741/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Pan-polymorph STING agonist

STING agonist C53 is an activator of human STING (EC50 = 185 nM). It promotes the oligomerization and activation of human STING through a mechanism orthogonal to that of cGAMP. STING agonist C53 increases activity of STING variants in HEK293T cells (107%, 92% and 92% reporter activity against H232, R232 and HAQ, respectively). It induces phosphorylation of STING variants in human tumor cell lines and PBMCs. STING agonist C53 is active at human but not mouse STING. Orally bioavailable.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-[(2-Chloro-6-fluorophenyl)methyl]-2,3-dihydro-3,3-dimethyl-2-oxo-N-[(2,4,6-trifluorophenyl)methyl]-1 H-indole-6-carboxamide
CAS 2259624-71-8
Quantity 10 mg
Target STING Agonists
Molecular Formula C25H19ClF4N2O2
Purity 0.98
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.