missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ STIM2 Polyclonal Antibody

Description
A suggested positive control is A-20 cell lysate. PA5-20373 can be used with blocking peptide PEP-0490.
In T lymphocytes, the sole pathway for Ca++ entry following antigen-receptor binding is through store-operated Ca++-release-activated Ca++ (CRAC) channels. These channels are made up of the pore-forming subunit ORAI1 and the stromal interaction molecule 1 (STIM1), a protein that functions as a Ca++ sensor and activates the CRAC channels, migrating to the plasma membrane from endoplasmic reticulum (ER)-like sites which act as the Ca++ store. A related molecule, STIM2, acts to inhibit the STIM1-mediated store-operated Ca++ entry, and can form complexes with STIM1, suggesting they may play a coordinated role in controlling Ca++ entry. At least three isoforms of STIM2 are known to exist.
Specifications
Specifications
| Antigen | STIM2 |
| Applications | Immunohistochemistry, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | STIM2 |
| Gene Accession No. | P83093, Q9P246 |
| Gene Alias | Kiaa1482; OTTHUMP00000123457; OTTHUMP00000123458; OTTHUMP00000216198; Stim2; Stromal interaction molecule 2 |
| Gene Symbols | STIM2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?