Learn More
Invitrogen™ STIM1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596668
Description
Immunogen sequence: RQRVAPKPPQ MSRAADEALN AMTSNGSHRL IEGVHPGSLV EKLPDSPALA KKALLALNHG LDKAHSLMEL SPSAPPGGSP HLDSSRSHSP SSPDPDTPSP VGDSRALQAS RNTRIPHLAG KKAVAEEDNG SIGEETDSSP GRKKFPLKIF KKPLKK; Positive Samples: LO2, Jurkat, BxPC-3, HeLa, Mouse brain, Mouse heart, Mouse kidney; Cellular Location: Cell membrane, Cytoplasm, Endoplasmic reticulum membrane, Single-pass type I membrane protein, cytoskeleton.
STIM1 (stromal interacting molecule; also known as GOK) acts as a sensor of calcium depletion within the endoplasmic reticulum and transduces the signal to Orai1, the presumptive CRAC channel at the plasma membrane. Following decrease of luminal calcium concentration, STIM1 oligomerizes and induces Orai1 to enable entry of extracellular calcium into the cytoplasm. However, the precise mechanism of STIM1-Orai1 interaction has not been elucidated yet. Many questions also remain to be solved around STIM1 functional distribution. It turns out that STIM1 associates with growing ends of microtubules and is involved in endoplasmic reticulum tubule extension.
Specifications
| STIM1 | |
| Polyclonal | |
| Unconjugated | |
| STIM1 | |
| D11S4896E; GOK; IMD10; RGD:1306831}; SIM; Stim1; stim1 {ECO:0000312; STIM1L; STRMK; Stromal interaction molecule 1; stromal interaction molecule 1 precursor; TAM; TAM1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 20866, 361618, 6786 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 0.75 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| P70302, P84903, Q13586 | |
| STIM1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 530-685 of human STIM1 (NP_0031472). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.