missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Stilbamidine Diisethionate, LoGiCal
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
Stilbamidine Diisethionate
Specifications
Specifications
| Chemical Name or Material | Stilbamidine Diisethionate |
| CAS | 140-59-0 |
| Analyte or Component Names | Stilbamidine Diisethionate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C16 H16 N4 . 2 C2 H6 O4 S |
| Certifications/Compliance | ISO 9001 |
| Synonym | Ethanesulfonic acid, 2-hydroxy-, compd. with 4,4'-(1,2-ethenediyl)bis[benzenecarboximidamide] (2:1) (9 CI), 4,4'-Stilbenedicarboxamidine, diisethionate (6 CI), Ethanesulfonic acid, 2-hydroxy-, compd. with 4,4'-stilbenedicarboxamidine (2:1) (8 CI), Ethanesulfonic acid, 2-hydroxy-, compd. with 4,4'-stilbenedicarboxamidine (7 CI), 4,4'-Stilbenedicarboxamidine, bis(2-hydroxyethanesulfonate) (8 CI), 4,4'-Stilbenedicarboxamidine di(beta-hydroxyethanesulfonate), 4,4'-Stilbenedicarboxamidine di(beta-hydroxyethanesulfonate), 4,4'-Stilbenedicarboxamidine di(beta-hydroxyethanesulfonate), 4,4'-Stilbenedicarboxamidine di(beta-hydroxyethanesulfonate), 4,4'-Stilbenedicarboxamidine di(beta-hydroxyethanesulfonate), 4,4'-Stilbenedicarboxamidine di(beta-hydroxyethanesulfonate), 4,4'-Stilbenedicarboxamidine di(beta-hydroxyethanesulfonate), 4,4'-Stilbenedicarboxamidine di(beta-hydroxyethanesulfonate), 4,4'-Stilbenedicarboxamidine di(beta-hydroxyethanesulfonate) |
| InChI Formula | InChI=1 S/C16H16N4.2C2H6O4S/c17-15(18)13-7-3-11(4-8-13)1-2-12-5-9-14(10-6-12)16(19)20;2*3-1-2-7(4,5)6/h1-10 H,(H3,17,18)(H3,19,20);2*3 H,1-2H2,(H,4,5,6) |
| SMILES | NC(=N)c1ccc(C=Cc2ccc(cc2)C(=N)N)cc1.OCCS(=O)(=O)O.OCCS(=O)(=O)O |
| IUPAC Name | 4-[2-(4-carbamimidoylphenyl)ethenyl]benzenecarboximidamide;2-hydroxyethanesulfonic acid |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction