Learn More
STIL Mouse anti-Human, Unconjugated, Clone: 1E12C1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66876-1-IG-150UL
This item is not returnable.
View return policy
Description
The STIL antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human STIL. This antibody recognizes human antigen. The STIL antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| STIL | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human |
| Western Blot, Immunohistochemistry, ELISA | |
| 1E12C1 | |
| Unconjugated | |
| 150 μL | |
| 6491 | |
| IgG1 |