missing translation for 'onlineSavingsMsg'
Learn More

Stigmasterol, TRC

Product Code. 17980085
Change view
Click to view available options
Quantity:
50 mg
1 g
5 g
Unit Size:
1g
50mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17980085 1 g 1g
17970075 5 g 5g
17938594 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17980085 Supplier Toronto Research Chemicals Supplier No. TRCS6867501G
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 83-48-7
Molecular Formula C29 H48 O
Molecular Weight (g/mol) 412.69
Synonym Stigmasta-5,22-dien-3-ol, (3β,22E)-, Stigmasta-5,22-dien-3β-ol (8CI), (24S)-24-Ethylcholesta-5,22-dien-3β-ol, (24S)-5,22-Stigmastadien-3β-ol, (24S)-Stigmast-5,22-dien-3β-ol, (3β,22E)-Stigmasta-5,22-dien-3-ol, 24-Ethyl-5,22-cholestadien-3β-ol, 24β-Ethyl-5,22-cholestadien-3β-ol, NSC 8095, Stigmasta-5,22(E)-dien-3β-ol, Stigmasterin, Stigmasterol, δ5,22-Stigmastadien-3β-ol, δ5-Stigmasterol, β-Stigmasterol
IUPAC Name (3S,8S,9S,10R,13R,14S,17R)-17-[(E,2R,5S)-5-ethyl-6-methylhept-3-en-2-yl]-10,13-dimethyl-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-ol
SMILES CC[C@H](\C=C\[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C)C(C)C

Specifications

Quantity 1 g
InChI Formula InChI=1S/C29H48O/c1-7-21(19(2)3)9-8-20(4)25-12-13-26-24-11-10-22-18-23(30)14-16-28(22,5)27(24)15-17-29(25,26)6/h8-10,19-21,23-27,30H,7,11-18H2,1-6H3/b9-8+/t20-,21-,23+,24+,25-,26+,27+,28+,29-/m1/s1
Formula Weight 412.3705
Percent Purity >95%
Chemical Name or Material Stigmasterol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.