missing translation for 'onlineSavingsMsg'
Learn More

Stigmasterol, MedChemExpress

Product Code. 16438996
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16438996 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16438996 Supplier Medchem Express Supplier No. HYN0131
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Stigmasterol is a plant sterol which has been focused on the cholesterol-lowering activity and is valued as an anti-stiffness factor in the therapy of rheumatic diseases.

Stigmasterol is a plant sterol which has been focused on the cholesterol-lowering activity and is valued as an anti-stiffness factor in the therapy of rheumatic diseases.

Specifications

Chemical Name or Material Stigmasterol
CAS 83-48-7
Color White
Purity Grade Notes Research
Molecular Formula C29H48O
Quantity 100 mg
Synonym Stigmasterin
Solubility Information Acetone : 6.67 mg/mL (16.16 mM; Need ultrasonic) ∣DMF : 1 mg/mL (2.42 mM; ultrasonic and warming and heat to 60°C) ∣Ethanol : < 1 mg/mL (insoluble) ∣MEOH : < 1 mg/mL (insoluble) ∣DMSO : < 1 mg/mL (insoluble or slightly soluble) ∣H2O : < 0.1 mg/mL (insoluble) ∣1M NaOH : < 1 mg/mL (insoluble)
SMILES C[C@H](/C=C/[C@@H](CC)C(C)C)[C@H]1CC[C@@]2([H])[C@]3([H])CC=C4C[C@@H](O)CC[C@]4(C)[C@@]3([H])CC[C@]12C
Molecular Weight (g/mol) 412.69
Formula Weight 412.69
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.