missing translation for 'onlineSavingsMsg'
Learn More

Stigmasterol 3-Acetate, TRC

Product Code. 30418247
Change view
Click to view available options
Quantity:
500 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30418247 5 g
30417646 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30418247 Supplier Toronto Research Chemicals Supplier No. TRCS6867555G
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4651-48-3
Molecular Formula C31H50O2
Molecular Weight (g/mol) 454.73
Synonym (3beta,22E)-Stigmasta-5,22-dien-3-ol Acetate, 3beta-Acetoxystigmasta-5,22-diene, NSC 8109, Stigmasteryl Acetate, Stigmasta-5,22-dien-3beta-ol Acetate, (3beta,22E)-Stigmasta-5,22-dien-3beta-ol 3-Acetate
IUPAC Name [(3S,8S,9S,10R,13R,14S,17R)-17-[(E,2R,5S)-5-ethyl-6-methylhept-3-en-2-yl]-10,13-dimethyl-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-yl] acetate
SMILES CC[C@H](\C=C\[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@H](CC[C@]4(C)[C@H]3CC[C@]12C)OC(=O)C)C(C)C

Specifications

Quantity 5 g
InChI Formula InChI=1S/C31H50O2/c1-8-23(20(2)3)10-9-21(4)27-13-14-28-26-12-11-24-19-25(33-22(5)32)15-17-30(24,6)29(26)16-18-31(27,28)7/h9-11,20-21,23,25-29H,8,12-19H2,1-7H3/b10-9+/t21-,23-,25+,26+,27-,28+,29+,30+,31-/m1/s1
Formula Weight 454.3811
Chemical Name or Material Stigmasterol 3-Acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.