missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STI1 Rabbit anti-Human, Mouse, Rat, Clone: 4S8L6, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-15246-100UL
This item is not returnable.
View return policy
Description
STI1 Monoclonal antibody specifically detects STI1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| STI1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 4S8L6 | |
| Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin | |
| Hop, HOPSTI1L, Hsc70/Hsp90-organizing protein, Hsp70/Hsp90-organizing protein, IEF-SSP-3521, NY-REN-11 antigen, Renal carcinoma antigen NY-REN-11, STI1P60, stress-induced-phosphoprotein 1, Transformation-sensitive protein IEF SSP 3521 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 2-98 of human STI1 (P31948). | |
| 100 μg | |
| Core ESC Like Genes, Stem Cell Markers | |
| 10963 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction